Zinn(II)-Tetrafluorborat, 50 % w/w, wässrige Lösung, Thermo Scientific Chemicals
Zinn(II)-Tetrafluorborat, 50 % w/w, wässrige Lösung, Thermo Scientific Chemicals
Zinn(II)-Tetrafluorborat, 50 % w/w, wässrige Lösung, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinn(II)-Tetrafluorborat, 50 % w/w, wässrige Lösung, Thermo Scientific Chemicals

CAS: 13814-97-6 | B2F8Sn | 292.32 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 kg
3 x 1 kg
Katalognummer 089669.A1
auch als 089669-A1 bezeichnet
Preis (EUR)
136,00
Each
Menge:
1 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
136,00
Each
Chemikalien-Kennzeichnungen
CAS13814-97-6
IUPAC Nameλ²-tin(2+) bis(tetrafluoroboranuide)
Molecular FormulaB2F8Sn
InChI KeyJPZSNSDMGTYJIW-UHFFFAOYSA-N
SMILES[Sn++].F[B-](F)(F)F.F[B-](F)(F)F
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Colorless to pale yellow
FormLiquid
Assay (unspecified)48.0-53.0%
Tin(II) tetrafluoroborate is used in electroplating. It is also used for plating bath tinned copper, tin and tin alloy. Further, it acts as a catalyst for various organic reactions. It is also employed in metal plating. In addition to this, it is used as a petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only