alpha-Amino-gamma-butyrolactone hydrobromide, 99%
alpha-Amino-gamma-butyrolactone hydrobromide, 99%
alpha-Amino-gamma-butyrolactone hydrobromide, 99%
alpha-Amino-gamma-butyrolactone hydrobromide, 99%
Thermo Scientific Chemicals

alpha-Amino-gamma-butyrolactone hydrobromide, 99%

CAS: 6305-38-0 | C4H7NO2·BrH | 182.02 g/mol
数量:
2.5 g
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 103290100
価格(JPY)
-
見積もりを依頼する
数量:
10 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6305-38-0
IUPAC Namehydrogen 3-aminooxolan-2-one bromide
Molecular FormulaC4H8BrNO2
InChI KeyMKLNTBLOABOJFZ-UHFFFAOYNA-N
SMILES[H+].[Br-].NC1CCOC1=O
さらに表示
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only