6-Aminopenicillanic acid, 96%
6-Aminopenicillanic acid, 96%
6-Aminopenicillanic acid, 96%
Thermo Scientific Chemicals

6-Aminopenicillanic acid, 96%

CAS: 551-16-6 | C8H12N2O3S | 216.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 104250100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS551-16-6
IUPAC Name(2S,5R,6R)-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular FormulaC8H12N2O3S
InChI KeyNGHVIOIJCVXTGV-ALEPSDHESA-N
SMILESCC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point197°C to 203°C
Appearance (Color)White to cream
Infrared spectrumConforms
HPLC>=95 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only