3-Aminopyridine, 99%
3-Aminopyridine, 99%
3-Aminopyridine, 99%
Thermo Scientific Chemicals

3-Aminopyridine, 99%

CAS: 462-08-8 | C5H6N2 | 94.12 g/mol
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 104565000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS462-08-8
IUPAC Namepyridin-3-amine
Molecular FormulaC5H6N2
InChI KeyCUYKNJBYIJFRCU-UHFFFAOYSA-N
SMILESNC1=CC=CN=C1
さらに表示
Appearance (Form)Crystals or flakes or crystalline mass
Appearance (Color)Off-white to brown
Infrared spectrumConforms
Melting point60°C to 63°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only