Antipyrine, 99%
Antipyrine, 99%
Antipyrine, 99%
Thermo Scientific Chemicals

Antipyrine, 99%

CAS: 60-80-0 | C11H12N2O | 188.23 g/mol
製品番号(カタログ番号) 104975000
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS60-80-0
IUPAC Name1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Molecular FormulaC11H12N2O
InChI KeyVEQOALNAAJBPNY-UHFFFAOYSA-N
SMILESCN1N(C(=O)C=C1C)C1=CC=CC=C1
さらに表示
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
Titration Iodimetric>=98.5 %
Melting point109°C to 113°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only