1-Bromo-3-phenylpropane, 98%
1-Bromo-3-phenylpropane, 98%
1-Bromo-3-phenylpropane, 98%
Thermo Scientific Chemicals

1-Bromo-3-phenylpropane, 98%

CAS: 637-59-2 | C9H11Br | 199.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 107290050
Price (EUR)
23,70
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
23,70
Each
Chemical Identifiers
CAS637-59-2
IUPAC Name(3-bromopropyl)benzene
Molecular FormulaC9H11Br
InChI KeyXMZQWZJMTBCUFT-UHFFFAOYSA-N
SMILESBrCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.5450 to 1.5470 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only