4-tert-Butylphenol, 97%
4-tert-Butylphenol, 97%
4-tert-Butylphenol, 97%
Thermo Scientific Chemicals

4-tert-Butylphenol, 97%

CAS: 98-54-4 | C10H14O | 150.22 g/mol
数量:
5 g
1 kg
2.5 kg
製品番号(カタログ番号) 108000025
価格(JPY)
-
数量:
2.5 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS98-54-4
IUPAC Name4-tert-butylphenol
Molecular FormulaC10H14O
InChI KeyQHPQWRBYOIRBIT-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(O)C=C1
さらに表示
Appearance (Color)White to light beige
Infrared spectrumConforms
GC>=96.0 %
Water=<0.5 % (K.F.)
Appearance (Form)Flakes or pastilles
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only