n-Butyl sulfide, 99%, pure
n-Butyl sulfide, 99%, pure
n-Butyl sulfide, 99%, pure
Thermo Scientific Chemicals

n-Butyl sulfide, 99%, pure

CAS: 544-40-1 | C8H18S | 146.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 108015000
Price (USD)
200.65
Special offer
Online exclusive
Ends: 28-Dec-2026
236.00
Save 35.35
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
200.65
Special offer
Online exclusive
Ends: 28-Dec-2026
236.00
Save 35.35
Each
Chemical Identifiers
CAS544-40-1
IUPAC Name1-(butylsulfanyl)butane
Molecular FormulaC8H18S
InChI KeyHTIRHQRTDBPHNZ-UHFFFAOYSA-N
SMILESCCCCSCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4520 to 1.4540 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only