4-Chloroaniline, 98%
4-Chloroaniline, 98%
4-Chloroaniline, 98%
Thermo Scientific Chemicals

4-Chloroaniline, 98%

CAS: 106-47-8 | C6H6ClN | 127.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 108591000
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.40
Save 4.75
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.40
Save 4.75
Each
Chemical Identifiers
CAS106-47-8
IUPAC Name4-chloroaniline
Molecular FormulaC6H6ClN
InChI KeyQSNSCYSYFYORTR-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to brown to purple
Melting point68°C to 72°C
AppearanceMay darken on storage
GC>=97.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only