B-27™ Supplement (50X), minus antioxidants
Gibco™

B-27™ Supplement (50X), minus antioxidants

A variation of the most cited neuronal cell culture supplement, B-27 Supplement Minus Antioxidants (AO) is a customized version ofRead more
Have Questions?
Catalog NumberQuantity
1088903810 mL
Catalog number 10889038
Price (KRW)
221,000
Online offer
Ends: 30-Sep-2026
245,000
Save 24,000 (10%)
Each
Quantity:
10 mL
Customize this product
Price (KRW)
221,000
Online offer
Ends: 30-Sep-2026
245,000
Save 24,000 (10%)
Each
A variation of the most cited neuronal cell culture supplement, B-27 Supplement Minus Antioxidants (AO) is a customized version of the original B-27 Supplement without five antioxidants (vitamin E, vitamin E acetate, superoxide dismutase, catalase, and glutathione). B-27 Supplement Minus AO is a serum-free supplement designed for studies of the role of free-radicals in aging, toxicity, apoptosis, and chronic neurologic diseases. B-27 Supplement Minus AO is provided as a 50X liquid and is intended for use with Neurobasal Medium or Neurobasal-A Medium for neuronal cell culture without the need for an astrocyte feeder layer.

B-27 Supplement is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of B-27 Supplement and Neurobasal media.

For established protocols using B-27 Supplement, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeNeural Cells
Concentration50 X
Culture TypeMammalian Cell Culture
Product LineB-27
Quantity10 mL
Serum LevelSerum-Free
Shelf Life12 Months
Shipping ConditionDry Ice
SourceSerum-Free
Tested ForEndotoxin, Performance, Sterility, Mycoplasma
ClassificationSerum-free
FormLiquid
Product TypeCell Culture Supplement
SterilitySterile
pH6 to 8
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -20°C) and protect from light.

Frequently asked questions (FAQs)

Can B-27 supplement be filtered?

Yes, the protein content in this product is high enough so filtering through a low protein binding filter as a 50X or 1X in solution should not be a problem.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can Neurobasal Medium alone or Neurobasal Medium supplemented with B-27 supplement be frozen to extend product shelf life?

No, it is not recommended to freeze media due to the potential of precipitates forming upon thaw. Inorganic salts and amino acids in the formulation may come out of solution when exposed to temperature fluctuations. These precipitates will not go back into solution easily once formed.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer B-27 supplements with and without insulin?

Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I thaw B-27 Supplements in a 37 degrees C water bath or incubator?

We do not recommend thawing at 37 degrees C. Best results are observed upon thawing at room temperature for approximately 2 hrs or overnight at 2-8 degrees C.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How do I thaw B-27 Supplements?

We recommend thawing it at 4 degrees C overnight or at room temperature for 2 hours. We don't recommend thawing in a water bath or incubator.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.