3-Chlorophenol, 99%
3-Chlorophenol, 99%
3-Chlorophenol, 99%
Thermo Scientific Chemicals

3-Chlorophenol, 99%

CAS: 108-43-0 | C6H5ClO | 128.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number 109811000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-43-0
IUPAC Name3-chlorophenol
Molecular FormulaC6H5ClO
InChI KeyHORNXRXVQWOLPJ-UHFFFAOYSA-N
SMILESOC1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Refractive index1.5650 to 1.5670 (20°C, 589 nm)
Appearance (Form)Liquid after melting
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only