Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Integrin beta 1 (CD29) Antibodies

          Invitrogen

          CD29 (Integrin beta 1) Monoclonal Antibody (eBioHMb1-1 (HMb1-1)), FITC, eBioscience™

          Advanced Verification
          69 Published Figures
          87 References
          View all (116) Integrin beta 1 (CD29) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD29 (Integrin beta 1) Monoclonal Antibody (eBioHMb1-1 (HMb1-1)), FITC, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (69)
          • Advanced Verification (1)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 71

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD29 (Integrin beta 1) Antibody (11-0291-80) in Flow

          Staining of C57Bl/6 bone marrow cells with Anti-Human/Mouse CD45R (B220) APC (Product # 17-0452-82) and 0.5 µg of Armenian Hamster IgG Isotype Control FITC (Product # 11-4888-81) (left) or 0.5 µg of Anti-Mouse/Rat CD29 (Integrin beta 1) FITC (right). Cells in the small scatter population were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Flow Cytometry (Flow)
          CD29 (Integrin beta 1) Antibody in Immunocytochemistry (ICC/IF)
          CD29 (Integrin beta 1) Antibody in Immunocytochemistry (ICC/IF)
          CD29 (Integrin beta 1) Antibody

          Product Details

          11-0291-80

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          1 µg/test
          View 81 publications 81 publications
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Human, Mouse, Rabbit, Rat

          Host/Isotype

          Armenian hamster / IgG

          Recommended Isotype Control

          Armenian Hamster IgG Isotype Control (eBio299Arm), FITC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBioHMb1-1 (HMb1-1)

          Conjugate

          FITC FITC FITC
          • Unconjugated
          • APC 
          • APC-eFluor 780
          • Biotin 
          • eFluor 450
          • Functional Grade
          • NovaFluor Blue 660-120S
          • NovaFluor Red 685
          • NovaFluor Red 700
          • NovaFluor Red 755
          • NovaFluor Yellow 660
          • NovaFluor Yellow 700
          • PE 
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 702
          • Request custom conjugation

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2572448

          Product Specific Information

          Description: The eBioHMb1-1 monoclonal antibody reacts with mouse and rat CD29 (integrin beta 1), a 110-120 kDa member of the beta integrin family expressed by leukocytes, endothelial, smooth muscle and epithelial cells. CD29 binds non-covalently with the alpha integrins CD49a-f to form the VLA-1 through VLA-6 complexes, as well as with CD51. These alpha-beta integrin heterodimers are capable of mediating a variety of cellular responses including adhesion, trafficking, proliferaton and differentiation. All integrins which include CD29 bind to extracellular matrix proteins including collagen, laminin, fibronectin and vitronectin, whereas some CD29-containing integrins can also interact with cellular receptors such as VCAM-1 and MadCAM-1.

          Applications Reported: This eBioHMb1-1 (HMb1-1) antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This eBioHMb1-1 (HMb1-1) antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD29, also known as ITGB1 or Integrin Subunit Beta 1, is a 110 kDa cell surface glycoprotein widely expressed by various cells, including all leukocytes. It forms non-covalently linked heterodimers with at least six different alpha integrins (CD49a-f), creating VLA-1 through VLA-6 complexes, as well as with CD51. These alpha-beta integrin heterodimers mediate a range of cellular responses, including adhesion, trafficking, proliferation, and differentiation. CD29-containing integrins bind to extracellular matrix proteins such as collagen, laminin, fibronectin, and vitronectin. Some integrins also interact with cellular receptors like VCAM-1 and MadCAM-1. CD29 plays a crucial role in immune cell trafficking from blood to tissue, particularly during leukocyte emigration and extravascular migration. It is involved in regulating tissue damage and disease symptoms related to inflammatory bowel disease. In the presence of anti-tumor drugs, decreased expression of ITGB1 correlates with multidrug resistance in tumor cells. ITGB1-dependent mechanisms enhance cytokine secretion in human airway smooth muscle in response to IL-1beta, facilitated by fibronectin and type I collagen. More than eight beta subunits with numerous splice variant isoforms have been identified in mammals, including two major forms: beta1A and beta1D. Beta1A is present in all tissues except cardiac and skeletal muscle, which express the beta1D variant.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: Beta OL; Beta oligodendroglia; CD29; Fibronectin receptor subunit beta; integrin beta 1 (fibronectin receptor beta); Integrin beta-1; Integrin beta1; integrin VLA-4 subunit beta; ITBG1D; VLA-4 subunit beta

          View more View less

          Gene Aliases: 4633401G24Rik; AA409975; AA960159; CD29; ENSMUSG00000051907; Fnrb; Gm9863; gpIIa; Itgb1

          View more View less

          UniProt ID: (Mouse) P09055, (Rat) P49134

          View more View less

          Entrez Gene ID: (Mouse) 16412, (Rat) 24511

          View more View less

          Function(s)
          glycoprotein binding fibronectin binding protease binding actin binding receptor activity receptor binding integrin binding protein binding collagen binding kinase binding protein kinase binding protein domain specific binding protein complex binding peptide binding laminin binding metal ion binding protein heterodimerization activity cell adhesion molecule binding alpha-actinin binding collagen binding involved in cell-matrix adhesion integrin
          Process(es)
          G1/S transition of mitotic cell cycle in utero embryonic development cell fate specification tissue homeostasis cell migration involved in sprouting angiogenesis cellular calcium ion homeostasis cell adhesion leukocyte cell-cell adhesion cell-matrix adhesion calcium-independent cell-matrix adhesion transforming growth factor beta receptor signaling pathway integrin-mediated signaling pathway regulation of G-protein coupled receptor protein signaling pathway positive regulation of cell proliferation negative regulation of cell proliferation germ cell migration visual learning regulation of collagen catabolic process positive regulation of cell-substrate adhesion positive regulation of neuron projection development cell migration formation of radial glial scaffolds positive regulation of cell migration neuron projection development negative regulation of cell projection organization cell-substrate adhesion protein transport within lipid bilayer heterotypic cell-cell adhesion negative regulation of Rho protein signal transduction positive regulation of apoptotic process stress fiber assembly positive regulation of MAPK cascade sarcomere organization negative regulation of cell differentiation negative regulation of neuron differentiation positive regulation of neuron differentiation positive regulation of endocytosis axon extension cardiac muscle tissue development dendrite morphogenesis positive regulation of peptidyl-tyrosine phosphorylation leukocyte tethering or rolling regulation of cell cycle cardiac muscle cell differentiation bicellular tight junction assembly positive regulation of establishment of protein localization to plasma membrane negative regulation of anoikis response to activity response to nutrient levels cell-cell adhesion mediated by integrin response to gonadotropin response to drug mesodermal cell differentiation maternal process involved in female pregnancy cellular response to mechanical stimulus cellular response to vitamin D cellular response to ionizing radiation response to transforming growth factor beta
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline