Opti-MEM™ I Reduced Serum Medium, no phenol red
Gibco™

Opti-MEM™ I Reduced Serum Medium, no phenol red

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in cell growth rate or morphology.
Have Questions?
Catalog NumberQuantity
110580211 x 500 mL
Catalog number 11058021
Price (USD)
83.25
Each
Quantity:
1 x 500 mL
Customize this product
Price (USD)
83.25
Each

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in cell growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I Reduced-Serum Medium modifications for a range of cell culture applications.

This Opti-MEM™ is modified as follows:

With: L-glutamine

Without: Phenol Red

The complete formulation is confidential. For more information, please contact Technical Services.

Using Opti-MEM™ Reduced-Serum Medium

Opti-MEM™ I medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineSp2, AE-1, CHO, BHK-21, and HEK
Cell TypeMammalian Cells
Concentration1X
Culture EnvironmentCO2
For Use With (Application)Mammalian Cell Culture, Lipid-Based Transfection
FormatBottle
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco, Opti-MEM
Product TypeOpti-MEM I
Quantity1 x 500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionAmbient
ClassificationHuman Origin
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelReduced Serum
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutamine
Without AdditivesNo Phenol Red, No FBS
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

Do you offer an alternative for Opti-MEM I Reduced Serum Medium (Cat. No. 31985070)?

Opti-MEM I Reduced Serum Medium, no phenol red (Cat. No. 11058021) would be the closest alternative. Both offerings contain L-glutamine. For more a stabilized L-glutamine option, Opti-MEM I Reduced Serum Medium, GlutaMAX Supplement (Cat. No. 51985034) is the recommended alternative. All Opti-MEM offerings have demonstrated performance for both reduced-serum cell culture and lipid-based transfection.

Is there any other difference between Opti-MEM I Reduced Serum Medium and Opti-MEM I Reduced Serum Medium, no phenol red besides the presence/absence of phenol red?

The formulation of Opti-MEM I Reduced Serum Medium and that of Opti-MEM I Reduced Serum Medium, no phenol red is the same except for the presence of phenol red in the former medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Where can I find the osmolality for my lot of Opti-MEM I Reduced Serum Medium?

We do provide osmolality information on the certificate of analysis. All lots of Opti-MEM I Reduced Serum Medium will meet the osmolality specification of ≥280 to ≤320 mOsm/kg.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I aliquot and freeze Opti-MEM I Reduced Serum Medium to conserve it longer? If so, for how long can it be stored?

Sorry, we don't have data to show that frozen Opti-MEM I Reduced Serum Medium has a longer shelf life. Actually, we recommend storing this medium at 2-8 degrees C in the dark. We do not recommend freezing the medium because it may precipitate.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I noticed that Opti-MEM I Reduced Serum Medium does not contain any serum. Why do you then refer to it as "Reduced Serum Medium"?

Opti-MEM I Reduced Serum Medium is a serum-free medium. The reason why it is called “reduced-serum medium” is because you only need to supplement this medium with 1-5% FBS instead of the standard 10% FBS.

Opti-MEM I Reduced Serum Medium can be used as is (without serum) for transfection of cells. However, for long-term cell culture, you need to add serum to the medium to keep cells healthy.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (2)

Citations & References
Abstract
L-phenylalanine and NPS R-467 synergistically potentiate the function of the extracellular calcium-sensing receptor through distinct sites.
Authors: Zhang Zaixiang; Jiang Yongfeng; Quinn Stephen J; Krapcho Karen; Nemeth Edward F; Bai Mei;
Journal:J Biol Chem
PubMed ID:12114500
The extracellular calcium (Ca(2+)(o))-sensing receptor (CaR) can be potentiated by allosteric activators including calcimimetics and l-amino acids. In this study, we found that many mutations had differential effects on the functional modulation of the CaR by these two allosteric activators, supporting the idea that these modulators act through distinct sites. ... More
Mutation of residues 423 (Met/Ile), 444 (Thr/Met), and 506 (Asn/Ser) confer cholesteryl esterase activity on rat lung carboxylesterase. Ser-506 is required for activation by cAMP-dependent protein kinase.
Authors: Wallace T J; Kodsi E M; Langston T B; Gergis M R; Grogan W M;
Journal:J Biol Chem
PubMed ID:11429416
Site-directed mutagenesis is used to identify amino acid residues that dictate reported differences in substrate specificity between rat hepatic neutral cytosolic cholesteryl ester hydrolase (hncCEH) and rat lung carboxylesterase (LCE), proteins differing by only 4 residues in their primary sequences. Beginning with LCE, the substitution Met(423) --> Ile(423) alone or ... More