Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement
Gibco™

Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
51985034500 mL
5198509110 x 500 mL
카탈로그 번호 51985034
제품 가격(KRW)
85,000
온라인 행사
Ends: 30-Sep-2026
94,000
할인액 9,000 (10%)
Each
수량:
500 mL
Customize this product
제품 가격(KRW)
85,000
온라인 행사
Ends: 30-Sep-2026
94,000
할인액 9,000 (10%)
Each

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I modifications for a range of cell culture applications.


This Opti-MEM™ I is modified as follows:
With
• GlutaMAX™
• Phenol Red


The complete formulation is confidential. For more information, please contact Technical Services.

Using Opti-MEM™ I Medium
Opti-MEM™ I Reduced-Serum Medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
사양
세포주Sp2, AE-1, CHO, BHK-21, and HEK
세포 유형Primary Fibroblasts
농도1X
제조 품질cGMP-compliant under the ISO 13485 standard
제품라인Gibco, GlutaMAX, Opti-MEM
제품 유형Opti-MEM I
수량500 mL
유통 기한18 Months From Date of Manufacture
배송 조건Room Temperature
분류Animal Origin
형태Liquid
멸균Sterile-filtered
첨가제 포함GlutaMAX, Phenol Red
Unit SizeEach
구성 및 보관
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture

자주 묻는 질문(FAQ)

Is there any other difference between Opti-MEM I Reduced Serum Medium and Opti-MEM I Reduced Serum Medium, no phenol red besides the presence/absence of phenol red?

The formulation of Opti-MEM I Reduced Serum Medium and that of Opti-MEM I Reduced Serum Medium, no phenol red is the same except for the presence of phenol red in the former medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Where can I find the osmolality for my lot of Opti-MEM I Reduced Serum Medium?

We do provide osmolality information on the certificate of analysis. All lots of Opti-MEM I Reduced Serum Medium will meet the osmolality specification of ≥280 to ≤320 mOsm/kg.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I aliquot and freeze Opti-MEM I Reduced Serum Medium to conserve it longer? If so, for how long can it be stored?

Sorry, we don't have data to show that frozen Opti-MEM I Reduced Serum Medium has a longer shelf life. Actually, we recommend storing this medium at 2-8 degrees C in the dark. We do not recommend freezing the medium because it may precipitate.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I noticed that Opti-MEM I Reduced Serum Medium does not contain any serum. Why do you then refer to it as "Reduced Serum Medium"?

Opti-MEM I Reduced Serum Medium is a serum-free medium. The reason why it is called “reduced-serum medium” is because you only need to supplement this medium with 1-5% FBS instead of the standard 10% FBS.

Opti-MEM I Reduced Serum Medium can be used as is (without serum) for transfection of cells. However, for long-term cell culture, you need to add serum to the medium to keep cells healthy.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

인용 및 참조 문헌 (11)

인용 및 참조 문헌
Abstract
Integrin-linked kinase regulates inducible nitric oxide synthase and cyclooxygenase-2 expression in an NF-kappa B-dependent manner.
Authors: Tan Clara; Mui Alice; Dedhar Shoukat;
Journal:J Biol Chem
PubMed ID:11724787
'Nitric oxide (NO) and prostaglandins are produced as a result of the stimulation of inducible nitric oxide synthase (iNOS) and cyclooxygenase-2, respectively, in response to cytokines or lipopolysaccharide (LPS). We demonstrate that the activity of integrin-linked kinase (ILK) is stimulated by LPS activation in J774 macrophages. Inhibition of ILK activity ... More
Doxorubicin induces apoptosis and CD95 gene expression in human primary endothelial cells through a p53-dependent mechanism.
Authors: Lorenzo Elisa; Ruiz-Ruiz Carmen; Quesada Antonio Jesús; Hernández Gabriela; Rodríguez Antonio; López-Rivas Abelardo; Redondo Juan Miguel;
Journal:J Biol Chem
PubMed ID:11779855
'Regulation of the homeostasis of vascular endothelium is critical for the processes of vascular remodeling and angiogenesis under physiological and pathological conditions. Here we show that doxorubicin (Dox), a drug used in antitumor therapy, triggered a marked accumulation of p53 and induced CD95 gene expression and apoptosis in proliferating human ... More
Structural and energetic characteristics of the heparin-binding site in antithrombotic protein C.
Authors:Friedrich U, Blom AM, Dahlbäck B, Villoutreix BO,
Journal:J Biol Chem
PubMed ID:11316800
'Human activated protein C (APC) is a key component of a natural anticoagulant system that regulates blood coagulation. In vivo, the catalytic activity of APC is regulated by two serpins, alpha1-antitrypsin and the protein C inhibitor (PCI), the inhibition by the latter being stimulated by heparin. We have identified a ... More
Phospholipase C-gamma is required for agonist-induced Ca2+ entry.
Authors: Patterson Randen L; van Rossum Damian B; Ford Diana L; Hurt Kenneth J; Bae Sun Sik; Suh Pann Ghill; Kurosaki Tomohiro; Snyder Solomon H; Gill Donald L;
Journal:Cell
PubMed ID:12437926
'We report here that PLC-gamma isoforms are required for agonist-induced Ca2+ entry (ACE). Overexpressed wild-type PLC-gamma1 or a lipase-inactive mutant PLC-gamma1 each augmented ACE in PC12 cells, while a deletion mutant lacking the region containing the SH3 domain of PLC-gamma1 was ineffective. RNA interference to deplete either PLC-gamma1 or PLC-gamma2 ... More
Hydrogen peroxide stimulates c-Src-mediated big mitogen-activated protein kinase 1 (BMK1) and the MEF2C signaling pathway in PC12 cells: potential role in cell survival following oxidative insults.
Authors: Suzaki Yuki; Yoshizumi Masanori; Kagami Shoji; Koyama A Hajime; Taketani Yutaka; Houchi Hitoshi; Tsuchiya Koichiro; Takeda Eiji; Tamaki Toshiaki;
Journal:J Biol Chem
PubMed ID:11782488
'Reactive oxygen species, generated by reduction-oxidation (redox) reactions, have been recognized as one of the major mediators of ischemia and reperfusion injury in the brain. Reactive oxygen species-induced cerebral events are attributable, in part, to the change in intracellular signaling molecules including mitogen-activated protein (MAP) kinases. Big MAP kinase 1 ... More