Cyanoacetic acid, 97%
Cyanoacetic acid, 97%
Cyanoacetic acid, 97%
Thermo Scientific Chemicals

Cyanoacetic acid, 97%

CAS: 372-09-8 | C3H3NO2 | 85.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 110670050
Price (USD)
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS372-09-8
IUPAC Name2-cyanoacetic acid
Molecular FormulaC3H3NO2
InChI KeyMLIREBYILWEBDM-UHFFFAOYSA-N
SMILESOC(=O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Titration with NaOH>=96.0 %
Appearance (Form)Adhering crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only