Cyclopropyl methyl ketone, 98%
Cyclopropyl methyl ketone, 98%
Cyclopropyl methyl ketone, 98%
Thermo Scientific Chemicals

Cyclopropyl methyl ketone, 98%

CAS: 765-43-5 | C5H8O | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 111710250
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.10
Save 7.45
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.10
Save 7.45
Each
Chemical Identifiers
CAS765-43-5
IUPAC Name1-cyclopropylethan-1-one
Molecular FormulaC5H8O
InChI KeyHVCFCNAITDHQFX-UHFFFAOYSA-N
SMILESCC(=O)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Water=<0.2 %
Appearance (Form)Liquid
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only