3,5-Diaminobenzoic acid, 98%
3,5-Diaminobenzoic acid, 98%
3,5-Diaminobenzoic acid, 98%
Thermo Scientific Chemicals

3,5-Diaminobenzoic acid, 98%

CAS: 535-87-5 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 112115000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS535-87-5
IUPAC Name5-carboxybenzene-1,3-bis(aminium) dichloride
Molecular FormulaC7H10Cl2N2O2
InChI KeyIGPLRBRBTUNCRT-UHFFFAOYSA-N
SMILES[Cl-].[Cl-].[NH3+]C1=CC(=CC([NH3+])=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
Appearance (Form)Powder
Infrared spectrumConforms
Titration with TBAH97.5 to 102.5 %
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only