2,6-Dichlorotoluene, 99+%
2,6-Dichlorotoluene, 99+%
2,6-Dichlorotoluene, 99+%
Thermo Scientific Chemicals

2,6-Dichlorotoluene, 99+%

CAS: 118-69-4 | C7H6Cl2 | 161.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 113791000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS118-69-4
IUPAC Name1,3-dichloro-2-methylbenzene
Molecular FormulaC7H6Cl2
InChI KeyDMEDNTFWIHCBRK-UHFFFAOYSA-N
SMILESCC1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.5500 to 1.5520 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only