Leibovitz's L-15 Medium, 500 mL - FAQs
View additional product information for Leibovitz's L-15 Medium - FAQs (11415049)
1 product FAQs found
What is the osmolality of Leibovitz's L-15 Medium?
We do provide osmolality information on the certificate of analysis. All lots of Leibovitz's L-15 Medium will meet the osmolality specification of 300-340 mOsm/kg.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.