3,5-Dimethylbenzoic acid, 99%
3,5-Dimethylbenzoic acid, 99%
3,5-Dimethylbenzoic acid, 99%
3,5-Dimethylbenzoic acid, 99%
Thermo Scientific Chemicals

3,5-Dimethylbenzoic acid, 99%

CAS: 499-06-9 | C9H10O2 | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 116001000
Price (TWD)
-
Request A Quote
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS499-06-9
IUPAC Name3,5-dimethylbenzoic acid
Molecular FormulaC9H10O2
InChI KeyUMVOQQDNEYOJOK-UHFFFAOYSA-N
SMILESCC1=CC(=CC(C)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH98.5 to 101.5 %
Appearance (Color)White to yellow
Melting point169°C to 172°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only