2,3-Dimethylbutane, 98+%
2,3-Dimethylbutane, 98+%
2,3-Dimethylbutane, 98+%
Thermo Scientific Chemicals

2,3-Dimethylbutane, 98+%

CAS: 79-29-8 | C6H14 | 86.18 g/mol
数量:
100 mL
1 L
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 116101000
価格(JPY)
-
数量:
100 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS79-29-8
IUPAC Name2,3-dimethylbutane
Molecular FormulaC6H14
InChI KeyZFFMLCVRJBZUDZ-UHFFFAOYSA-N
SMILESCC(C)C(C)C
さらに表示
Refractive index1.3740 to 1.3760 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=98.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only