2,6-Dimethylphenol, 99%
2,6-Dimethylphenol, 99%
2,6-Dimethylphenol, 99%
Thermo Scientific Chemicals

2,6-Dimethylphenol, 99%

CAS: 576-26-1 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 116570025
Price (USD)
152.65
Special offer
Online exclusive
Ends: 28-Dec-2026
179.00
Save 26.35
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
152.65
Special offer
Online exclusive
Ends: 28-Dec-2026
179.00
Save 26.35
Each
Chemical Identifiers
CAS576-26-1
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.05 %
Melting point45°C to 48°C
Appearance (Color)Colorless to yellow-brown
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only