SuperScript™ III First-Strand Synthesis SuperMix for qRT-PCR
Invitrogen™

SuperScript™ III First-Strand Synthesis SuperMix for qRT-PCR

SuperScript III First-Strand Synthesis SuperMix for qRT-PCR provides the high-temperature capability of SuperScript III Reverse Transcriptase in an optimized SuperMix format for the synthesis of first-strand cDNA for use in real-time quantitative RT-PCR (qRT-PCR).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
11752250250 Reactions
1175205050 Reactions
Catalog number 11752250
Price (USD)
3,230.65
Online exclusive
3,590.00
Save 359.35
Each
No. of Reactions:
250 Reactions
Request bulk or custom format
Price (USD)
3,230.65
Online exclusive
3,590.00
Save 359.35
Each

SuperScript III First-Strand Synthesis SuperMix for qRT-PCR provides the high-temperature capability of SuperScript III Reverse Transcriptase in an optimized SuperMix format for the synthesis of first-strand cDNA for use in real-time quantitative RT-PCR (qRT-PCR). The simple, time-saving reaction set-up uses just two tubes: a 2X reaction mix and an enzyme mix.

Enzyme mix
SuperScript III Reverse Transcriptase, included in the RT enzyme mix, is a version of M-MLV RT that has been engineered to reduce RNase H activity and provide increased thermal stability. The enzyme can be used to synthesize cDNA at a temperature range of 42–60°C, providing increased specificity, higher yields of cDNA, and more full-length product than other reverse transcriptases. Because SuperScript III RT is not significantly inhibited by ribosomal and transfer RNA, it can be used to synthesize cDNA from total RNA. RNaseOUT Recombinant Ribonuclease Inhibitor, also included in the enzyme mix, is an RNase inhibitor protein that safeguards against the degradation of target RNA due to ribonuclease contamination of the RNA preparation.

Reaction mix
The 2X RT reaction mix includes oligo(dT)20, random hexamers, MgCl2, and dNTPs in a buffer formulation that has been optimized for qRT-PCR. E. coli RNase H is provided as a separate tube in the kit to remove the RNA template from the cDNA:RNA hybrid molecule after first-strand synthesis. This has been shown to increase sensitivity in qRT-PCR.

Using SuperScript III First-Strand Synthesis SuperMix
This SuperMix formulation can be used to quantify fewer than 10 copies of a target gene in qRT-PCR, with a broad dynamic range that supports accurate quantification of high-copy mRNA from up to 1 μg of total RNA. Reagents are provided for 50 or 250 RT reactions of 20 μL each.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Final Product TypeFirst-Strand cDNA
FormatSuper Mix
No. of Reactions250 Reactions
Optimal Reaction Temperature50°C
Quantity250 rxns
Reaction FormatMaster Mix
Reagent TypeReverse Transcription
Reverse TranscriptaseSuperScript III
Shipping ConditionDry Ice
Starting MaterialRNA
TechniqueReverse Transcription
Detection MethodqPCR
For Use With (Application)Real Time PCR (qPCR)
GC-Rich PCR PerformanceHigh
Reaction Speed30 min.
Unit SizeEach
Contents & Storage

• 2X RT Reaction Mix (2 x 1.25 mL)
• RT Enzyme Mix (500 μL)
• E. coli RNase (250 μL)

Sufficient reagents for 250 reactions at 20 μL reaction volumes.
Store at –20°C.

Frequently asked questions (FAQs)

How can I remove genomic DNA contamination from my sample prior to performing RT-PCR?

We recommend using ezDNase (Cat. No. 11766051). ezDNase Enzyme's high specificity for double-stranded DNA enables efficient and fast genomic DNA removal without reduction in the quality or quantity of RNA. ezDNase Enzyme is heat-labile and so can be easily deactivated by heat treatment at moderate temperature (55 degrees C). These features make ezDNase Enzyme an excellent choice for genomic DNA removal prior to reverse transcription reactions.

How much RNA should be employed for first-strand cDNA synthesis?

The amount of RNA template for a cDNA synthesis is highly flexible and depends upon the amount of sample available and an individual's need. In general, 1 µg total RNA is used in a typical 20-µL RT reaction.

Find additional tips, troubleshooting help, and resources within ourReverse Transcription and RACE Support Center.

Should I treat the cDNA with RNase H prior to downstream processing?

RNase H treatment is not always necessary. Many PCR reactions work without it. However, for cDNA synthesized with RNase H-deficient reverse transcriptases (like SuperScript II, III, and IV), RNA/cDNA hybrids—especially GC-rich ones—may not denature well, reducing PCR sensitivity. RNase H treatment can help in such cases. Additionally, RNase H treatment is beneficial for cloning larger fragments.

What percentage of RNA is converted to cDNA when performing reverse transcription?

This depends highly on the quality of the sample. mRNA itself makes up 1-5% of total RNA. Depending on the primer and enzyme used, reverse transcription can covert >70% of that into cDNA.

Find additional tips, troubleshooting help, and resources within our Reverse Transcription and RACE Support Center.

I'm setting up my RT reaction and am trying to decide whether I should use random primers, oligo(dT) primer, gene-specific primer, or oligo(dT)/random mix primers. What would you suggest?

Random primers are the best choice for degraded RNA, RNA with heavy secondary structure, non-polyadenylated RNA, or prokaryotic RNA. It is recommended only for two-step RT-PCR, and typically gives the highest yields, although the cDNA may not necessarily be full length. Oligo(dT) primers are good to use when trying to recover full-length cDNA from 2-step RT-PCR. The reaction is influenced by secondary structure and RNA quality. Gene specific primers should be used for very specific, mainly one-step RT-PCR reactions.

Find additional tips, troubleshooting help, and resources within our Reverse Transcription and RACE Support Center.

Citations & References (3)

Citations & References
Abstract
Simvastatin downregulates the expression of hepcidin and erythropoietin in HepG2 cells.
Authors:Chang CC, Chiu PF, Chen HL, Chang TL, Chang YJ, Huang CH
Journal:Hemodial Int
PubMed ID:22716163
'Statin therapy may improve responsiveness to erythropoietin-stimulating agents in patients with end-stage renal disease. Although statins increase hepatic iron uptake and storage capacity in cholestatic rats, the underlying mechanisms are unclear. Therefore, we examined the effects of a statin (simvastatin) on the expression of hepcidin, erythropoietin receptor (EPOR) and erythropoietin ... More
TDAG51 is an ERK signaling target that opposes ERK-mediated HME16C mammary epithelial cell transformation.
Authors:Oberst MD, Beberman SJ, Zhao L, Yin JJ, Ward Y, Kelly K,
Journal:BMC Cancer
PubMed ID:18597688
'INTRODUCTION: Signaling downstream of Ras is mediated by three major pathways, Raf/ERK, phosphatidylinositol 3 kinase (PI3K), and Ral guanine nucleotide exchange factor (RalGEF). Ras signal transduction pathways play an important role in breast cancer progression, as evidenced by the frequent over-expression of the Ras-activating epidermal growth factor receptors EGFR and ... More
Reprogramming of mouse fibroblasts into cardiomyocyte-like cells in vitro.
Authors:Qian L, Berry EC, Fu JD, Ieda M, Srivastava D,
Journal:
PubMed ID:23722259
Cardiac fibroblasts can be reprogrammed to cardiomyocyte-like cells by the introduction of three transcription factors: Gata4, Mef2c and Tbx5 (collectively referred to here as GMT). Resident cardiac fibroblasts can be converted in vivo into induced cardiomyocyte-like cells (iCMs) that closely resemble endogenous cardiomyocytes and electrically integrate with the host myocardium. ... More