SuperScript™ IV VILO™ Master Mix, Each - FAQs
SuperScript™ IV VILO™ Master Mix, Each - FAQs
View additional product information for SuperScript™ IV VILO™ Master Mix - FAQs (11766050, 11756500, 11766500, 11756050)
27 product FAQs found
Which Reverse Transcription kit can I use for FFPE samples?
For FFPE samples we recommend SSIV VILO Master Mix. This mix works better with degraded RNA.
Find additional tips, troubleshooting help, and resources within our Reverse Transcription and RACE Support Center.
In your SuperScript IV RT protocols, there is no ezDNase inactivation step. Will active ezDNase affect RNA or the RT reaction?
The Invitrogen ezDNase Enzyme is a novel DNase that is highly specific for double-stranded DNA. It has no activity on single-stranded DNA in RT reactions (primers or probes), or on RNA. The enzyme is also thermolabile—it is inactivated quickly at temperatures typical for the SuperScript IV RT reaction (e.g., 50°C). The additional inactivation step is therefore not required in RT-qPCR applications.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Which SuperScript IV RT format do you recommend for real-time PCR applications?
For RT-qPCR applications we recommend using the Invitrogen SuperScript IV VILO Master Mix (Cat. No. 11756050). The cDNA synthesis reaction setup with this master mix requires fewer pipetting steps and therefore reduces variation in the data. SuperScript IV RT, as a component of the master mix, offers the highest efficiency of cDNA synthesis step compared to competitors’ products.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Are there any significant changes in the SuperScript IV RT protocol compared to the SuperScript III RT protocol?
The only change is that the incubation time for the reverse transcription reaction has been reduced from 50 minutes to 10 minutes. All the other parameters and steps are the same.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Can I get comparable cDNA yield and length using the SuperScript IV RT 10-minute protocol as when using the 50-minute protocol for SuperScript III RT?
When compared with SuperScript III RT (and other manufacturers’ RTs) in a synthesis reaction for a 9 kb cDNA, SuperScript IV RT performed successful synthesis in just 10 minutes and did so with comparable (or improved) yield (as shown by gel band density).
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Do you offer a one-step RT-qPCR kit containing SuperScript IV RT?
We have not created a one-step RT-qPCR kit containing SuperScript IV RT for real-time PCR. However, it is included in end-point one-step RT-PCR kits: https://thermofisher.com/ssiv-onestep
We also offer a two-step RT-qPCR kit containing SuperScript IV RT: https://www.thermofisher.com/vilo
For one-step RT-qPCR, we offer a kit with SuperScript III RT: https://www.thermofisher.com/order/catalog/product/117320200
What control do you recommend using with the Oncomine TCR Beta-LR Assay?
We recommend using T Cell Leukemia (Jurkat) Total RNA (Cat. No. AM7858) as a control. T cell Leukemia (Jurkat) Total RNA is derived from a cell line consisting of a single T cell clonotype. Running the Oncomine TCR Beta-LR Assay on Jurkat Total RNA should enable detection of a single clonotype.
Are there any BED files for the Oncomine TCR Beta-LR Assay?
Unlike other AmpliSeq panels, there are no BED files and no reference is needed. All downstream analysis takes place in Ion Reporter Software.
Can I quantitate the Oncomine TCR Beta-LR Assay library with the Ion Library Equalizer Kit?
We have tested the Ion Library Equalizer Kit with this kit for library quantitation and it does function, but manually balancing library input into the Ion Chef instrument is preferred.
What version of Torrent Suite Software and Ion Reporter Software can be used with the Oncomine TCR Beta-LR Assay?
Torrent Suite Software 5.6 and Ion Reporter Software 5.6 are needed to generate reports. In Torrent Suite Software 5.6, two Planned Run templates are provided for use with the Oncomine TCR Beta-LR Assay.
Can I run the Oncomine TCR Beta-LR Assay on the Ion PGM System?
The Oncomine TCR Beta-LR Assay has been developed and verified for use on the Ion GeneStudio S5 Series System. The assay can be used with the Ion PGM System, however, this instrument configuration is not supported.
What are the components of the Oncomine TCR Beta-LR Assay?
Here are the components of the Oncomine TCR Beta-LR Assay:
- Oncomine TCR Beta‑LR Panel
- Ion AmpliSeq Library Kit Plus
- Ion Select Barcode Adapters
What is the concentration of the primer pool in the Oncomine TCR Beta-LR Assay?
The primer pool is at 5X concentration.
What are the amplicon read lengths in the Oncomine TCR Beta-LR Assay?
The Oncomine TCR Beta-LR Assay enables long-read RNA sequencing (up to 400 bp amplicon length) for complete characterization of the T cell receptor Beta (TCR Beta) chain, including all three complementarity-determining regions (CDR1, 2, and 3) of the variable gene. This enables allele-specific interrogation of TCR Beta to potentially predict drug-mediated adverse events.
How many primer pools does the Oncomine TCR Beta-LR Assay kit contain?
The primers are provided as a single pool.
With the Oncomine TCR Beta-LR Assay, how many samples can I multiplex on each chip?
Up to 16 samples can be multiplexed on an Ion 530 chip, up to 4 samples can be multiplexed on an Ion 520 chip, and up to 4 samples can be multiplexed on an Ion 318 chip.
What research sample types are compatible with the Oncomine TCR Beta-LR Assay?
A variety of research sample types including fresh-frozen tissue, whole blood, and sorted T cells are compatible with the Oncomine TCR Beta-LR Assay.
Why are immunologists and immuno-oncologists interested in T cell repertoire sequencing research?
T cell repertoire sequencing helps:
- To characterize diversity and study features of T lymphocytes in blood and infiltrated tumors
- To study variable gene polymorphisms implicated in autoimmune disease or immune-mediated adverse events
- To study the production and function of therapeutic or engineered T cells
How many samples can I analyze with the Oncomine TCR Beta-LR Assay?
This kit is good for 24 samples.
What does the Oncomine TCR Beta-LR Assay measure?
The Oncomine TCR Beta-LR Assay is an RNA-based next-generation sequencing (NGS) research assay that enables the characterization of the T cell receptor Beta (TCR Beta) sequences including all three complementarity-determining regions (CDR1, 2, and 3) of the variable gene. The assay measures T cell repertoire diversity as well as clonal expansion, and allows for identification of allele-specific polymorphisms.
What is the difference between SuperScript IV RT and the RT enzyme used in SuperScript IV VILO Master Mix?
The SuperScript IV VILO Master Mix contains SuperScript IV RT, a ribonuclease inhibitor, and a helper protein. The helper protein helps to increase the efficiency of the reverse transcription reaction and thus improve cDNA yield. This master mix formulation allows for a simpler reaction setup with less pipetting and less variation between samples.
How many reactions of SuperScript IV VILO no RT Control are included with the SuperScript IV VILO Master Mix?
For the 50 reaction SKUs, the no RT control can be for 50 reactions. For the 500 reaction SKUs, the no RT control can be for 250 reactions, which is way more than enough for control reaction purpose.
Why did you use oligo (dT)18 plus random hexamer primers for SuperScript IV VILO Master Mix whereas the SuperScript VILO Master Mix contains only random hexamer primers?
This is to help ensure that the biases from using either oligo (dT)18 or random hexamer primers alone will be eliminated, although the data indicate that using random hexamers alone provides quantitation just as good as the combination of the two primers. Some scientists want the combination because they feel it could be better for data quality.
How does SuperScript IV VILO Master Mix perform with GC-rich RNA templates?
SuperScript IV VILO Master Mix has increased thermostability, allowing the RT incubation temperature to be raised to deal with the structural complexity often associated with GC-rich templates. Our tests show that compared to other cDNA synthesis kits or master mixes, the raised reaction temperature leads to minimal loss of cDNA synthesis efficiency for SuperScript IV VILO Master Mix, suggesting that it is the best choice if high reaction temperature is needed for cDNA synthesis from GC-rich templates.
What is the recommended reaction temperature for SuperScript IV VILO Master Mix?
The recommended reaction temperature for SuperScript IV VILO Master Mix is 50 degrees C. R&D has tested the stability of SSIV VILO at higher temperatures and it works very well even at 65 degrees C.
What are the key performance improvements of SuperScript IV VILO Master Mix over the SuperScript VILO Master Mix and SuperScript VILO kits?
SuperScript IV VILO Master Mix uses SuperScript IV Reverse Transcriptase (RT) in the optimized master mix formulation, whereas SuperScript VILO Master Mix and SuperScript VILO kits use SuperScript III Reverse Transcriptase. SuperScript IV RT has a significantly improved performance profile over SuperScript III RT in thermostability, sensitivity, yield, processivity, and the ability to synthesize cDNA efficiently from a wide variety of RNA samples, even those of suboptimal purity and integrity.
Which applications can SuperScript IV VILO Master Mix be used for?
SuperScript IV VILO Master Mix is an upgrade from SuperScript VILO Master Mix and cDNA Synthesis Kit. It is designed to be used for cDNA synthesis in the two-step RT-qPCR applications for gene expression analysis.