Ethyl vinyl ether, 99%, stabilized
Ethyl vinyl ether, 99%, stabilized
Ethyl vinyl ether, 99%, stabilized
Thermo Scientific Chemicals

Ethyl vinyl ether, 99%, stabilized

CAS: 109-92-2 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 119080010
Price (USD)
85.65
Online exclusive
95.20
Save 9.55
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
85.65
Online exclusive
95.20
Save 9.55
Each
Chemical Identifiers
CAS109-92-2
IUPAC Nameethoxyethene
Molecular FormulaC4H8O
InChI KeyFJKIXWOMBXYWOQ-UHFFFAOYSA-N
SMILESCCOC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless
Refractive index1.3760 to 1.3780 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only