2-Fluorobenzaldehyde, 97%
2-Fluorobenzaldehyde, 97%
2-Fluorobenzaldehyde, 97%
Thermo Scientific Chemicals

2-Fluorobenzaldehyde, 97%

CAS: 446-52-6 | C7H5FO | 124.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 119321000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS446-52-6
IUPAC Name2-fluorobenzaldehyde
Molecular FormulaC7H5FO
InChI KeyZWDVQMVZZYIAHO-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Refractive index1.5205 to 1.5225 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only