Hexanophenone, 98%
Hexanophenone, 98%
Hexanophenone, 98%
Thermo Scientific Chemicals

Hexanophenone, 98%

CAS: 942-92-7 | C12H16O | 176.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 120720050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS942-92-7
IUPAC Name1-phenylhexan-1-one
Molecular FormulaC12H16O
InChI KeyMAHPVQDVMLWUAG-UHFFFAOYSA-N
SMILESCCCCCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear light yellow
Appearance (Form)Liquid after melting
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only