Homopiperazine, 98%
Homopiperazine, 98%
Homopiperazine, 98%
Thermo Scientific Chemicals

Homopiperazine, 98%

CAS: 505-66-8 | C5H12N2 | 100.165 g/mol
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 120851000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS505-66-8
IUPAC Name1,4-diazepane
Molecular FormulaC5H12N2
InChI KeyFQUYSHZXSKYCSY-UHFFFAOYSA-N
SMILESC1CNCCNC1
さらに表示
Appearance (Color)White to light yellow
Infrared spectrumConforms
Water=<1 %
Appearance (Form)Crystalline mass
Melting point38°C to 42°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only