This 1X TE Buffer is a component of the PureLink™ 96 Plasmid Purification System, now offered separately. It is usedRead more
Have Questions?
Catalog Number
Quantity
12090015
100 mL
Catalog number 12090015
Price (USD)
118.65
Online exclusive
124.00
Save 5.35
Each
Quantity:
100 mL
Price (USD)
118.65
Online exclusive
124.00
Save 5.35
Each
This 1X TE Buffer is a component of the PureLink™ 96 Plasmid Purification System, now offered separately. It is used to resuspend the final purified plasmid pellet and contains very low EDTA, so it is compatible with sequencing and other enzymatic applications.
Composition: 10 mM Tris-HCl (pH 8.0) 0.1 mM EDTA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1 X
FormLiquid
Product TypeTE Buffer
Quantity100 mL
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.
Frequently asked questions (FAQs)
How do I download the Pathogen Interpretive Software?
Once you place your order, please contact our Instrument Service team at 1-800-955-6288 option 3, 1 (this kit is only available in North America). You will be given an access code to the TaqPath Monkeypox/Orthopox Virus DNA Kit eLearning where you’ll learn how to install, update, and download the Pathogen Interpretive Software package for the TaqPath Monkeypox/Orthopox Virus DNA Kit.
With the TaqPath Monkeypox/Orthopox Virus DNA Kit, what software should I use for data analysis?
Results are analyzed using the following software:
- Applied Biosystems Pathogen Interpretive Software v1.1
- SAE Administrator Console Dx v1.0 or v1.2 (for security and audit functions)
- Pathogen Interpretive Software 1.0.0 DAT (SAE Profile)
Please use the appropriate assay panel and template for your instrument:
- MPXOPX-EUA_QS5-9601_1.0.0.zip
- MPXOPX-EUA_QS5Dx-9602_1.0.0.zip
- MPXOPX-EUA_QS7F-384_1.0.0.zip
Note: These zip files are located in EducationConnect (www.thermofisher.com/educationconnect). The eLearning course needs to be completed to be granted access.
What is the instrument software configuration required to use the TaqPath Monkeypox/Orthopox Virus DNA Kit?
To obtain the correct analysis software or firmware version for your real-time PCR instrument, go to thermofisher.com/validatedfirmware, then select your instrument in the Real-Time PCR section.
Which instruments are authorized under the EUA for use with the TaqPath Monkeypox/Orthopox Virus DNA Kit?
Here are the instruments that can be used with the TaqPath Monkeypox/Orthopox Virus DNA Kit:
- Applied Biosystems QuantStudio 5 Real-Time PCR System (96-well, 0.1 mL block)
- Applied Biosystems QuantStudio 5 Dx Real-Time PCR System (96-well, 0.2 mL block)
- Applied Biosystems QuantStudio 7 Flex Real-Time PCR System (384-well block)
I would like to use the TaqPath Monkeypox/Orthopox Virus DNA Kit. Which extraction kit should I use to extract DNA from the sample?
Performance was established using the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit (Cat. No. A48383) in combination with the KingFisher Flex Magnetic Particle Processor with 96 Deep-Well Head and the KingFisher Flex 96 Deep-Well Heating Block (MVP_2Wash_400_Flex program). Other purification methods may lead to deviations in the expected result.