4-Hydroxy-6-methyl-2-pyrone, 98%
4-Hydroxy-6-methyl-2-pyrone, 98%
4-Hydroxy-6-methyl-2-pyrone, 98%
Thermo Scientific Chemicals

4-Hydroxy-6-methyl-2-pyrone, 98%

CAS: 675-10-5 | C6H6O3 | 126.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
12156025025 gGlass bottle
Catalog number 121560250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS675-10-5
IUPAC Name2-hydroxy-6-methyl-4H-pyran-4-one
Molecular FormulaC6H6O3
InChI KeyOOKCZXGEYPSNIM-UHFFFAOYSA-N
SMILESCC1=CC(=O)C=C(O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point186°C to 192°C
Appearance (Color)Off-white to brown
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only