3-Hydroxypyridine, 98%
3-Hydroxypyridine, 98%
3-Hydroxypyridine, 98%
Thermo Scientific Chemicals

3-Hydroxypyridine, 98%

CAS: 109-00-2 | C5H5NO | 95.1 g/mol
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 121855000
Price (JPY)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS109-00-2
IUPAC Namepyridin-3-ol
Molecular FormulaC5H5NO
InChI KeyGRFNBEZIAWKNCO-UHFFFAOYSA-N
SMILESOC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.2 %
Appearance (Color)Beige to brown
Melting point123°C to 130°C
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only