Isochroman, 99%
Isochroman, 99%
Isochroman, 99%
Thermo Scientific Chemicals

Isochroman, 99%

CAS: 493-05-0 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 122540250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS493-05-0
IUPAC Name3,4-dihydro-1H-2-benzopyran
Molecular FormulaC9H10O
InChI KeyHEBMCVBCEDMUOF-UHFFFAOYSA-N
SMILESC1CC2=CC=CC=C2CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5430 to 1.5460 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only