DMEM, low glucose, pyruvate, HEPES
Gibco™

DMEM, low glucose, pyruvate, HEPES

Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells.詳細を見る
製品番号(カタログ番号)数量
12320032500 mL
製品番号(カタログ番号) 12320032
価格(JPY)
5,000
Each
数量:
500 mL
Customize this product

Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This DMEM is modified as follows:
With
• Low Glucose
• L-glutamine
• Sodium Pyruvate
• HEPES
• Phenol Red



The complete formulation is available.

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.

DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
仕様
細胞株HeLa, 293, Cos-7, and PC-12
細胞タイプPrimary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
濃度1 X
製造品質cGMP-compliant under the ISO 13485 standard
製品ラインGibco
製品タイプDMEM (Dulbecco's Modified Eagle Medium)
数量500 mL
品質保持期間12 Months From Date of Manufacture
出荷条件Room Temperature
分類Animal Origin-free
形状Liquid
Serum LevelStandard Serum Supplementation
無菌性Sterile-filtered
添加剤ありLow Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate
Unit SizeEach
組成および保存条件
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

よくあるご質問(FAQ)

What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?

Manganese is not present in the formulation of our catalog DMEM media products.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.