FreeStyle™ 293 Expression Medium
Gibco™

FreeStyle™ 293 Expression Medium

An animal origin-free, serum-free, protein-free formulation, allowing for easier purification
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1233800220 L
123380181 L
123380266 x 1 L
1233800110 L
Catalog number 12338002
Price (USD)
2,925.65
Online exclusive
3,180.00
Save 254.35
Each
Quantity:
20 L
Price (USD)
2,925.65
Online exclusive
3,180.00
Save 254.35
Each

Gibco FreeStyle 293 Expression Medium is an animal origin–free, chemically defined, protein-free medium specifically developed to support the growth and transfection of 293-F cells in suspension without adaptation. The medium is complete and ready-to-use, with no supplementation required.

Features of FreeStyle 293 Expression Medium include:

  • Complete and ready-to-use format containing GlutaMAX supplement
  • Ability to support suspension growth and transfection of 293-F cells
  • Animal origin–free, protein-free and chemically defined formulation
  • Easy use with the FreeStyle 293 expression systems
  • Scalability in spinner flasks and bioreactors

Complete and ready-to-use format containing GlutaMAX supplement

FreeStyle 293 Expression Medium is ready to use. It does not require the addition of serum, glutamine, or surfactants. It contains GlutaMAX supplement to minimize toxic ammonia build-up and improve cell viability and growth in an easy-to-use format.

Ability to support suspension growth and transfection of 293-F cells

Gibco 293-F, 293-H, and FreeStyle 293-F cells in suspension can be cultured directly into FreeStyle 293 Expression Medium without adaptation. Cells adapted to other commercially available serum-free media can be subcultured directly into FreeStyle 293 Expression Medium, usually without any further adaptation. Cells usually require adaptation from serum-containing formulations. Cells cultured in FreeStyle 293 Expression Medium can be transfected using lipid based transfection reagents, such as 293fectin reagent (Cat. No. 12347019) and Freestyle MAX reagent (16447500).

Animal origin–free, protein-free and chemically defined formulation

FreeStyle 293 Expression Medium is animal origin–free, protein-free and chemically defined, allowing for easier purification of your protein of interest. Gibco chemically defined media contain no proteins, hydrolysates, or components of unknown composition.

Easy use as part of the FreeStyle 293 Expression System and FreeStyle MAX Expression System for protein production

The FreeStyle 293 Expression System (Cat. No. K900001) is a complete, suspension cell culture system for generating large amounts of mammalian recombinant protein (see figure). It combines FreeStyle 293 Expression Medium with 293fectin reagent, a cationic-lipid formulation designed to transfect suspension FreeStyle 293-F cells at high efficiency. The FreeStyle MAX Expression System is a breakthrough technology for rapid and high-yield mammalian protein production. It combines FreeStyle media, FreeStyle MAX Reagent, and either FreeStyle CHO-S cells or FreeStyle 293-F cells.

Scalability in spinner flasks and bioreactors

Protein production using FreeStyle 293 Expression Medium can be scaled up in spinner flasks or bioreactors. The appropriate spinner or impeller speed and seeding density should be optimized for each system. Depending on the impeller design and speed, it may be necessary to supplement the medium with additional Pluronic F-68 to avoid sheer stress in the culture.

Universal bag packaging

Gibco universal bags offer flexible packaging that allows you to integrate a single-use, disposable large volume media container with a wide variety of automated cell culture instruments, disposable bioreactor systems, and sterile connection processes, minimizing the need for costly customization.

Product use

Customers using FreeStyle 293 Expression Medium in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineFreeStyle™ 293-F Cells
Concentration1X
Culture EnvironmentCO2
Endotoxin LevelLow
Product LineFreestyle
Product TypeExpression Medium
Purity or Quality GradeCell Culture Grade
Quantity20 L
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture TypeSuspension Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile
With AdditivesGlutaMAX, Sodium Pyruvate, Sodium Bicarbonate
Without AdditivesNo Phenol Red
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

Can I load the FreeStyle 293 Expression Medium or FreeStyle CHO Expression Medium directly onto a nickel column for His-tagged protein purification?

With FreeStyle 293 Expression Medium, we would recommend harvesting the cells a little earlier to reduce any nonspecific binding by host cell proteins and then filtering the culture supernatant through a 0.2 or 0.1 µM membrane before loading onto the column. On the other hand, FreeStyle CHO Expression Medium contains EDTA that will strip the nickel column. The medium can be subjected to dialysis or buffer exchange prior to loading on the column, or the EDTA can be chelated by adding 1-3 mg/L NiSO4 or NiCl to the supernatant. Iron or cobalt salts can also be used.

Can FreeStyle 293-F cells grown in FreeStyle 293 expression medium be transfected with ExpiFectamine293 Reagent along with enhancers? Is so, will the transfection efficiency be higher than that with FreeStyle Max Reagent?

You should be able to use the ExpiFectamine293 Reagent for transfection of Freestyle 293-F cells grown in FreeStyle 293 Expression Medium; however, the enhancers will provide only a little boost in expression as they are designed to work with higher density cultures that FreeStyle 293 Expression Medium cannot support.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

Is it necessary to grow FreeStyle 293-F cells in 8% CO2 when using the FreeStyle 293 Expression Medium? If 8% CO2 cannot be arranged, can I supplement the media with an additional reagent?

Our recommendation of 8% CO2 for the optimal growth of Freestyle 293 cells in Freestyle 293 Expression Medium is based on the bicarbonate content in the media and growth conditions of the cells observed by R&D. That said, you may use lower concentrations of CO2 if necessary (5% is common) to grow these cells. You may observe slightly slower growth kinetics or lower cell concentrations, but functionally the entire system will still work.

Does FreeStyle 293 Expression Medium contain any components that would interfere with concentrating secreted proteins from HEK 293 cells via ammonium sulfate precipitation?

Yes, the FreeStyle 293 Expression Medium contains Pluronic F-68 and lipids that would interfere with ammonium sulfate precipitation. Dialysis or tangential flow filtration can be used to remove lipids and Pluronic F-68 from the medium. Then you can concentrate the medium with ammonium sulfate.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between FreeStyle 293 Expression Medium and FreeStyle F17 Expression Medium?

FreeStyle F17 Expression Medium was designed from a different base formulation than FreeStyle 293 Expression Medium. FreeStyle F17 medium shows equal or better performance than the FreeStye 293 medium for certain cell lines and proteins. It is capable of supporting the growth of 293 and CHO cells and offers a wider variety and selection of transient cell culture media options.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.