Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.
Thermo Scientific Chemicals

Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.

Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr., Quantity: 500mL, Packaging: Glass bottle, Colorless to Yellow, Molecular Weight: 259.47, Beilstein: 04,II,634, CAS: 2052-49-5, 108-88-3, 67-56-1, Density: 0.8500g/mL, EINECS Number: 218-147-6 | CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 123531000
Price (USD)
87.65
Online exclusive
97.70
Save 10.05
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
87.65
Online exclusive
97.70
Save 10.05
Each
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy colorless to yellow
Appearance (Form)Solution
Titration with HClO40.09 to 0.11 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only