4-Iodobenzoic acid, 98%
4-Iodobenzoic acid, 98%
4-Iodobenzoic acid, 98%
Thermo Scientific Chemicals

4-Iodobenzoic acid, 98%

CAS: 619-58-9 | C7H5IO2 | 248.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 124480100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS619-58-9
IUPAC Name4-iodobenzoic acid
Molecular FormulaC7H5IO2
InChI KeyGHICCUXQJBDNRN-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)Powder
HPLC>=97.5 %
Melting point270°C to 274°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only