3-Methoxyphenylacetic acid, 99.5%
3-Methoxyphenylacetic acid, 99.5%
3-Methoxyphenylacetic acid, 99.5%
Thermo Scientific Chemicals

3-Methoxyphenylacetic acid, 99.5%

CAS: 1798-09-0 | C9H10O3 | 166.18 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 126010050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1798-09-0
IUPAC Name2-(3-methoxyphenyl)acetate
Molecular FormulaC9H9O3
InChI KeyLEGPZHPSIPPYIO-UHFFFAOYSA-M
SMILESCOC1=CC=CC(CC([O-])=O)=C1
さらに表示
Appearance (Color)White to light beige
Appearance (Form)Powder and/or chunks
Melting point68°C to 72°C
Titration with NaOH>=99.4 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only