Methyl acrylate, 99%, stabilized
Methyl acrylate, 99%, stabilized
Methyl acrylate, 99%, stabilized
Thermo Scientific Chemicals

Methyl acrylate, 99%, stabilized

CAS: 96-33-3 | C4H6O2 | 86.09 g/mol
数量:
10 mL
500 mL
1 L
2.5 L
10 L
パッケージ:
すべて表示
Metal Drum
製品番号(カタログ番号) 126190101
価格(JPY)
-
数量:
10 L
パッケージ:
Metal Drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS96-33-3
IUPAC Namemethyl prop-2-enoate
Molecular FormulaC4H6O2
InChI KeyBAPJBEWLBFYGME-UHFFFAOYSA-N
SMILESCOC(=O)C=C
さらに表示
Appearance (Form)Clear liquid
Refractive index1.4010 to 1.4040 (20°C, 589 nm)
Color scale=<10 APHA
Infrared spectrumConforms
Water=<0.08 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only