2-(Methylthio)aniline, 97%
2-(Methylthio)aniline, 97%
2-(Methylthio)aniline, 97%
Acros Organics

2-(Methylthio)aniline, 97%

CAS: 2987-53-3 | C7H9NS | 139.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 127110250
HSN 29309099
Price (INR)
25,263.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
25,263.00
Each
Chemical Identifiers
CAS2987-53-3
IUPAC Name2-(methylsulfanyl)aniline
Molecular FormulaC7H9NS
InChI KeyWBRPQQSADOCKCH-UHFFFAOYSA-N
SMILESCSC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Light yellow to brown
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only