Mucinsäure, 98 %, Thermo Scientific Chemicals
Mucinsäure, 98 %, Thermo Scientific Chemicals
Mucinsäure, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mucinsäure, 98 %, Thermo Scientific Chemicals

CAS: 526-99-8 | C6H10O8 | 210.138 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 g
500 g
Verpackung:
Kunststoffflasche
Katalognummer 128041000
Preis (EUR)
48,90
Each
Menge:
100 g
Verpackung:
Kunststoffflasche
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
48,90
Each
Chemikalien-Kennzeichnungen
CAS526-99-8
IUPAC Name2,3,4,5-tetrahydroxyhexanedioic acid
Molecular FormulaC6H10O8
InChI KeyDSLZVSRJTYRBFB-UHFFFAOYNA-N
SMILESOC(C(O)C(O)C(O)=O)C(O)C(O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White to light beige
Titration excess NaOH>=97.5 %
Infrared spectrumConforms
Loss on drying=<0.5 %
Appearance (Form)Crystalline powder
Mehr anzeigen
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only