Picolinic acid, 99%
Picolinic acid, 99%
Picolinic acid, 99%
Thermo Scientific Chemicals

Picolinic acid, 99%

CAS: 98-98-6 | C6H5NO2 | 123.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Catalog number 131170025
Price (EUR)
703,00
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (EUR)
703,00
Each
Chemical Identifiers
CAS98-98-6
IUPAC Namepyridine-2-carboxylic acid
Molecular FormulaC6H5NO2
InChI KeySIOXPEMLGUPBBT-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH98.5 to 101.5 %
Appearance (Color)White to light yellow
Melting point136°C to 139°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only