3,5-Pyridinedicarboxylic acid, 98%
3,5-Pyridinedicarboxylic acid, 98%
3,5-Pyridinedicarboxylic acid, 98%
Thermo Scientific Chemicals

3,5-Pyridinedicarboxylic acid, 98%

CAS: 499-81-0 | C7H5NO4 | 167.12 g/mol
数量:
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 131900500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS499-81-0
IUPAC Namepyridine-3,5-dicarboxylic acid
Molecular FormulaC7H5NO4
InChI KeyMPFLRYZEEAQMLQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CN=C1)C(O)=O
さらに表示
Infrared spectrumConforms
Appearance (Form)Powder
Appearance (Color)White to off-white
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only