2-(Hydroxymethyl)pyridine, 98%
2-(Hydroxymethyl)pyridine, 98%
2-(Hydroxymethyl)pyridine, 98%
Thermo Scientific Chemicals

2-(Hydroxymethyl)pyridine, 98%

CAS: 586-98-1 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 131990250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS586-98-1
IUPAC Name(pyridin-2-yl)methanol
Molecular FormulaC6H7NO
InChI KeySHNUBALDGXWUJI-UHFFFAOYSA-N
SMILESOCC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Infrared spectrumConforms
Refractive index1.5410 to 1.5450 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only