Sodium hydroxide, 98%, extra pure, pellets
Sodium hydroxide, 98%, extra pure, pellets
Sodium hydroxide, 98%, extra pure, pellets
Thermo Scientific Chemicals

Sodium hydroxide, 98%, extra pure, pellets

CAS: 1310-73-2 | HNaO | 39.997 g/mol
数量:
2.5 kg
10 kg
25 kg
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 134070100
価格(JPY)
-
数量:
10 kg
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
さらに表示
Appearance (Form)Pellets
Sodium carbonate (Na2CO3)=<1.0 %
Iron (Fe)=<5 ppm
Nickel (Ni)=<5 ppm
Potassium (K)=<250 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only