1-Tetradecylamine, 98%
1-Tetradecylamine, 98%
1-Tetradecylamine, 98%
Thermo Scientific Chemicals

1-Tetradecylamine, 98%

CAS: 2016-42-4 | C14H31N | 213.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Packaging:
Glass bottle
Catalog number 138100100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2016-42-4
IUPAC Namehydrogen tetradecan-1-amine chloride
Molecular FormulaC14H32ClN
InChI KeyKKWUACQXLWHLCX-UHFFFAOYSA-N
SMILES[H+].[Cl-].CCCCCCCCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink
Titration with HClO4>=97.5 %
Appearance (Form)Crystals or crystalline solid
Infrared spectrumConforms
Melting point37°C to 42°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only