Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: GSSTQALTY LNALQHSIRL SGVEDHVKNF RPQCLVMTGA PNSRPALLHL VHDFTKNVGL MICGHVHMGP RRQAMKEMSI DQAKYQRWLI KNKMKAFYAP VHADDLREGA QYLMQAAGLG RMKPNTLVLG FKKDWLQADM RDVDMYINLF HDAFDIQYGV VVIRLKEGLD ISHLQGQEEL LSSQEKSPGT KDVVVSVEYS KKSDLDTSKP LSEKPITHKV EEEDGKTATQ PLLKKESKGP IVPLNVADQK LLEASTQFQK KQGKNTIDVW WLFDDGGLTL LIPYLLTTKK KWKDCKIRVF IGGKINRIDH DRRAMATLLS KFRIDFSDIM VLGDINTKPK KENIIAFEEI IEPYRLHEDD KEQDIADKMK EDEPWRITDN ELELYKTKFY EPC (759-1150 aa encoded by B C033003)
NKCC1 mediates sodium and chloride transport and reabsorption. NKCC1 is a membrane protein and is important in maintaining proper ionic balance and cell volume. NKCC1 is an electrically silent transporter system that mediates sodium and chloride reabsorption. Further, NKCC1 plays a vital role in the regulation of ionic balance and cell volume and is phosphorylated in response to DNA damage. Three transcript variants encoding two different isoforms have been found for NKCC1. Diseases associated with SLC12A2 include Basosquamous Carcinoma and Metatypical Basal Cell Carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basolateral Na-K-Cl symporter; BSC2; bumetanide-sensitive cotransporter type 2; bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1; Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; cation-chloride cotransporter; hNKCC1; membrane electrolyte transporter; mNKCC1; Na-K-2Cl cotransporter 1; protein phosphatase 1, regulatory subunit 141; sodium/potassium/chloride cotransporters; sodium/potassium/chloride transporters; solute carrier family 12 (sodium/potassium/chloride transporter), member 2; solute carrier family 12 (sodium/potassium/chloride transporters), member 2; Solute carrier family 12 member 2; solute carrier family 12, member 2
Gene Aliases: 9330166H04Rik; BSC; BSC-2; BSC2; CCC1; hNKCC1; KILQS; mBSC2; NKCC1; PPP1R141; SLC12A2; sy-ns
UniProt ID: (Human) P55011, (Mouse) P55012
Entrez Gene ID: (Human) 6558, (Rat) 83629, (Mouse) 20496
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support