Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • E-cadherin Antibodies

          Invitrogen

          CD324 (E-Cadherin) Monoclonal Antibody (DECMA-1), eBioscience™

          Advanced Verification
          45 Published Figures
          75 References
          View all (167) E-cadherin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD324 (E-Cadherin) Monoclonal Antibody (DECMA-1), eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (45)
          • Advanced Verification (1)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 48

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD324 (E-Cadherin) Antibody (14-3249-80) in ICC/IF

          Immunofluorescence analysis of E-cadherin was performed using 90% confluent log phase MCF7 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with CD324 (E-Cadherin) Monoclonal Antibody (DECMA-1) (Product # 14-3249-82) at 5 µg/mL in 0.1% BSA, incubated at 4 d... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Western Blot (WB)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Neutralization (Neu)
          CD324 (E-Cadherin) Antibody in Neutralization (Neu)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunocytochemistry (ICC/IF)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (IHC)
          CD324 (E-Cadherin) Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          CD324 (E-Cadherin) Antibody in Western Blot (WB)
          CD324 (E-Cadherin) Antibody in Western Blot (WB)
          CD324 (E-Cadherin) Antibody in Western Blot (WB)
          CD324 (E-Cadherin) Antibody in Western Blot (WB)
          CD324 (E-Cadherin) Antibody in Western Blot (WB)
          CD324 (E-Cadherin) Antibody in Western Blot (WB)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Flow Cytometry (Flow)
          CD324 (E-Cadherin) Antibody in Neutralization (Neu)
          CD324 (E-Cadherin) Antibody in Neutralization (Neu)
          CD324 (E-Cadherin) Antibody in Neutralization (Neu)
          CD324 (E-Cadherin) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          14-3249-80

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          View 11 publications 11 publications

          Immunohistochemistry (IHC)

          -
          View 22 publications 22 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 3 publications 3 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          View 20 publications 20 publications

          Flow Cytometry (Flow)

          Assay-Dependent
          View 11 publications 11 publications

          ELISA (ELISA)

          Assay-Dependent
          -

          Immunoprecipitation (IP)

          Assay-Dependent
          -

          Neutralization (Neu)

          -
          View 3 publications 3 publications

          Functional Assay (Functional)

          Assay-Dependent
          -

          Inhibition Assays (Inhibition)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse

          Published species

          Dog, Fruit fly, Human, Mouse

          Host/Isotype

          Rat / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          DECMA-1

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Biotin 
          • eFluor 660
          • Functional Grade
          • PerCP-eFluor 710
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1210459

          Product Specific Information

          Description: The monoclonal antibody DECMA-1 recognizes mouse, human and canine CD324 also known as E-cadherin (Epithelial cadherin) or uvomorulin. Like the other cadherin family members P and N cadherin, E-cadherin is a transmembrane glycoprotein involved in intercellular adhesion. These proteins share a common basic structure. The extracellular portions of the proteins are largely composed of repeating domains, each with two consensus Ca2+-binding motifs. The cytoplasmic domain interacts with a-, b-, and g-catenins and actinins. These catenins connect E-cadherin with the cytoskeleton.

          Expression is found in most epidermal cells including melanocytes and kerotinocytes. E-cadherin is localized at the intercellular boundaries of epithelial cells in several tissues, and is thought to play a role in maintenance of tissue integrity. Loss of E-cadherin function has been implicated in the progression of a variety of cancers.

          E-Cadherin protein is sensitive to trypsin treatment, so exposure to trypsin should be minimized or avoided.

          The monoclonal antibody DECMA-1 has been shown to have functional activity by disrupting adhesion in human, mouse and dog cells.

          Applications Reported: This DECMA-1 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, western blotting, immunohistochemical staining of frozen tissue sections, and ELISA. (Please use Functional Grade purified DECMA-1, Product # 16-3249-85 , in functional assays).

          Applications Tested: This DECMA-1 antibody has been tested by western blot analysis of EDTA treated MDCK cell line. This can be used at less than or equal to 2 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          E-Cadherin (epithelial cadherin) is a classical cadherin from the cadherin (alcium dependent adhesion protein) superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. E-cadherin plays a central role in the growth and development of cells by controlling tissue architecture, and maintenance of tissue integrity. In humans, E-cadherin is encoded by the CDH1 gene present on chromosome 16. Studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cadherin 1, E-cadherin (epithelial); cadherin 1, type 1, E-cadherin (epithelial); cadherin e; cadherin-E; calcium-dependent adhesion protein, epithelial; CAM 120/80; CDH1; cell-CAM 120/80; E-cadherin; E-cadherin 1; Ecadherin; epididymis secretory sperm binding protein; Epithelial cadherin; unnamed protein product; uvomorulin

          View more View less

          Gene Aliases: ARC-1; BCDS1; Cadherin-1; CD324; CDHE; E-cad; ECAD; L-CAM; LCAM; Um; UVO; Uvomorulin

          View more View less

          UniProt ID: (Human) P12830, (Dog) F1PAA9, (Mouse) P09803

          View more View less

          Entrez Gene ID: (Human) 999, (Dog) 442858, (Mouse) 12550

          View more View less

          Function(s)
          calcium ion binding protein binding beta-catenin binding ankyrin binding GTPase activating protein binding identical protein binding gamma-catenin binding cadherin binding cell adhesion molecule binding cell adhesion mediator activity protein tyrosine kinase binding cytoskeletal protein binding protein phosphatase binding protein domain specific binding alpha-catenin binding binding, bridging protein binding involved in cell-cell adhesion cadherin
          Process(es)
          cell morphogenesis desmosome assembly cell-cell junction assembly cell adhesion homophilic cell adhesion via plasma membrane adhesion molecules synapse assembly response to xenobiotic stimulus response to toxic substance regulation of gene expression calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules cell migration pituitary gland development negative regulation of cell-cell adhesion negative regulation of cell migration negative regulation of axon extension neuron projection development cell junction assembly adherens junction organization positive regulation of protein import into nucleus cell-cell adhesion mediated by cadherin positive regulation of transcription, DNA-templated cellular response to lithium ion response to heparin cellular response to indole-3-methanol protein localization to plasma membrane cell-cell adhesion regulation of protein catabolic process at postsynapse, modulating synaptic transmission response to Gram-positive bacterium positive regulation of cellular protein localization in utero embryonic development trophectodermal cell differentiation epithelial cell morphogenesis embryo implantation sensory perception of sound negative regulation of protein processing positive regulation of cell-cell adhesion actin cytoskeleton organization regulation of protein localization uterine epithelium development decidualization negative regulation of epithelial cell proliferation intestinal epithelial cell development salivary gland cavitation regulation of branching involved in salivary gland morphogenesis establishment of skin barrier bicellular tight junction assembly cellular response to amino acid stimulus negative regulation of canonical Wnt signaling pathway cochlea development negative regulation of protein localization to plasma membrane regulation of protein localization to cell surface regulation of neuron migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0