Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TIE2 (TEK) Antibodies

          Invitrogen

          CD202b (TIE2) Monoclonal Antibody (TEK4), eBioscience™

          24 Published Figures
          36 References
          View all (54) TIE2 (TEK) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD202b (TIE2) Monoclonal Antibody (TEK4), eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (24)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 25

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD202b (TIE2) Antibody (14-5987-82) in Flow

          Staining of mouse bone marrow with Anti-Mouse CD202b (TIE2) PE and Anti-Mouse Ly-6A/E (Sca-1) APC (Product # 17-5981-82) + Anti-Mouse CD117 (c-Kit) APC (Product # 17-1171-82). Total viable lineage negative cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          CD202b (TIE2) Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD202b (TIE2) Antibody in Immunocytochemistry (ICC/IF)
          CD202b (TIE2) Antibody in Immunohistochemistry (IHC)
          CD202b (TIE2) Antibody in Immunohistochemistry (IHC)
          CD202b (TIE2) Antibody in Immunohistochemistry (IHC)
          CD202b (TIE2) Antibody in Immunohistochemistry (IHC)
          CD202b (TIE2) Antibody in Immunohistochemistry (IHC)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)
          CD202b (TIE2) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          14-5987-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 11 publications 11 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 5 publications 5 publications

          Flow Cytometry (Flow)

          1 µg/test
          View 19 publications 19 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse, Pig

          Host/Isotype

          Rat / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          TEK4

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • APC 
          • Biotin 
          • Functional Grade
          • PE 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_467792

          Product Specific Information

          Description: The TEK4 monoclonal antibody reacts with mouse Tie-2, also known as CD202. A member of the tyrosine kinase receptor family, Tie-2 is expressed on endothelial and a subset of hematopoietic cells and is believed to play a role in both angiogenesis and hematopoiesis during development of the mouse embryo. In fetal liver and adult bone marrow, Tie-2 is expressed by a subpopulation of hematopoietic progenitor cells characterized as Lineage markers^-, c-Kit^+, Sca1^+ cells. Long-term multilineage repopulating cells were detected in Tie-2^+, Lineage^-, c-Kit^+, Sca1^+ cells but not in Tie-2^-, Lineage^-, c-Kit^+, Sca1^+ cells.

          Applications Reported: This TEK4 antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections.

          Applications Tested: The TEK4 antibody has been tested by blocking of staining with fluorochrome conjugated TEK4 on mouse bone marrow cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells. TEK signaling appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis, and defects in TEK are associated with inherited venous malformations. The ligand angiopoietin-1 binds to this receptor and mediates a signaling pathway that functions in embryonic vascular development. Immunoblotting showed that TIE2 expression was increased by thyroid-stimulating hormone and agents that increased intracellular cAMP. HSCs expressing the receptor tyrosine kinase TIE2 are quiescent and antiapoptotic and comprise a side population of HSCs that adhere to osteoblasts in the bone marrow niche.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD202b antigen; cell surface receptor homolog; EC 2.7.10; EC 2.7.10.1; endothelial tyrosine kinase; endothelial-specific receptor tyrosine kinase; p140 TEK; TEKT1; Tektin-1; tunica interna endothelial cell kinase; tyrosine kinase with Ig and EGF homology domains-2; tyrosine-protein kinase receptor TEK; tyrosine-protein kinase receptor TIE-2

          View more View less

          Gene Aliases: Cd202b; Hyk; STK1; Tie-2; Tie2

          View more View less

          UniProt ID: (Mouse) Q02858

          View more View less

          Entrez Gene ID: (Mouse) 21687

          View more View less

          Function(s)
          protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity protein binding ATP binding transmembrane receptor protein kinase activity growth factor binding signaling receptor activity identical protein binding
          Process(es)
          angiogenesis patterning of blood vessels vasculogenesis response to hypoxia positive regulation of protein phosphorylation endothelial cell proliferation regulation of endothelial cell proliferation sprouting angiogenesis positive regulation of cytokine production involved in immune response cell-matrix adhesion cell surface receptor signaling pathway transmembrane receptor protein tyrosine kinase signaling pathway heart development positive regulation of signal transduction positive regulation of endothelial cell migration negative regulation of angiogenesis peptidyl-tyrosine phosphorylation hemopoiesis regulation of cell migration positive regulation of vascular endothelial growth factor receptor signaling pathway cell-substrate adhesion response to retinoic acid regulation of establishment or maintenance of cell polarity positive regulation of peptidyl-serine phosphorylation substrate adhesion-dependent cell spreading positive regulation of protein import into nucleus negative regulation of apoptotic process positive regulation of MAPK cascade response to estrogen regulation of angiogenesis positive regulation of angiogenesis positive regulation of cell adhesion Tie signaling pathway branching morphogenesis of an epithelial tube positive regulation of multicellular organismal process positive regulation of focal adhesion assembly positive regulation of protein kinase B signaling heart trabecula formation positive regulation of ERK1 and ERK2 cascade cellular response to mechanical stimulus glomerulus vasculature development cell-cell adhesion regulation of NIK/NF-kappaB signaling regulation of intracellular signal transduction positive regulation of intracellular signal transduction regulation of endothelial cell apoptotic process negative regulation of endothelial cell apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0