Triphenylphosphine, 99%
Triphenylphosphine, 99%
Triphenylphosphine, 99%
Thermo Scientific Chemicals

Triphenylphosphine, 99%

CAS: 603-35-0 | C18H15P | 262.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 140420050
Price (USD)
547.65
Special offer
Online exclusive
Ends: 28-Dec-2026
644.00
Save 96.35
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
547.65
Special offer
Online exclusive
Ends: 28-Dec-2026
644.00
Save 96.35
Each
Chemical Identifiers
CAS603-35-0
IUPAC Nametriphenylphosphane
Molecular FormulaC18H15P
InChI KeyRIOQSEWOXXDEQQ-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
Melting point78.5°C to 81.5°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only