n-Undecane, 99%
n-Undecane, 99%
n-Undecane, 99%
Thermo Scientific Chemicals

n-Undecane, 99%

CAS: 1120-21-4 | C11H24 | 156.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
10 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 140665000
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS1120-21-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4160 to 1.4180 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only